HiTrap Columns, Capto SP ImpRes, Cytiva
Supplier: Cytiva
17-5468-51 17-5468-55
97067-776EA 218.96 USD
97067-776 97067-778
HiTrap Columns, Capto SP ImpRes, Cytiva
Chromatography Columns
HiTrap Capto SP ImpRes chromatography column is packed with a strong cation exchange modern resin for high resolution, small-scale protein purification.
  • Suitable for screening of selectivity, binding and elution conditions, and for small-scale purifications
  • Sulphonate (SP) strong cation exchanger
  • Optimized for intermediate and polishing protein purification
  • Convenient HiTrap format for easy connection to a syringe, peristaltic pump, or chromatography system such as an ÄKTA system
HiTrap Capto SP ImpRes column is prepacked with Capto SP ImpRes resin.More Product Information