VWR® TRIS Glycine Buffer
Supplier: VWR
M115-40L
97063-776EA 31.12 USD
97063-776 97063-780 97063-778 97063-782
VWR® TRIS Glycine Buffer
Buffers Electrophoresis Buffers Transfer Buffers
Following electrophoresis, proteins are frequently transferred from the polyacrylamide gel onto PVDF or nitrocellulose membranes for use in immunoblotting or sequencing
  • Convenience of liquid concentrates or powder blend
  • Consistent performance and quality
Although there are different methods by which to achieve transfer, the one most routinely used is electroblotting. Electroblotting utilizes conductive buffers that do not interfere with proteins binding to the membrane and can be performed in a vertical buffer tank or in a semi-dry blotting apparatus.