3,3',5,5'-Tetramethylbenzidine (TMB) membrane peroxidase substrate Plus, liquid, Biotechnology Grade 1 component

Supplier: VWR
Warning

Synonyms: TMB, 3,3',5,5'-Tetramethyl-[1,1'-biphenyl]-4,4'-diamine

K830-100ML
97063-666EA 164.33 USD
97063-666
3,3',5,5'-Tetramethylbenzidine (TMB) membrane peroxidase substrate Plus, liquid, Biotechnology Grade 1 component
3,3',5,5'-Tetramethylbenzidine (TMB)

Ready to use sensitive substrate for the detection of horseradish peroxidase (HRP) activity suitable for ELISA and solution assays. Offers faster signal, enhanced sensitivity and the lowest possible background.


  • Ready to use solution


Reaction of the substrate with HRP generates a free-radical cation that exhibits blue colour with an absorption maximum at 653 nm. Acidification of the reaction by addition of sulfuric acid yields a yellow-coloured diimine terminal oxidation product, which absorbs light at 450 nm.


TMB Plus liquid component substrate is optimised with TMB and hydrogen peroxide concentrations that allow a linear response to typical HRP concentrations utilised in immunologic assays.


Ordering information: Contains 2,08 mmol/l TMB, hydrogen peroxide, citric acid buffer pH 3,3 and additional proprietary, non toxic stabilisers.


Packaging: Packed in plastic bottle.

Formula: C₁₆H₂₀N₂
MW: 240.35 g/mol
MDL Number: MFCD00007748
CAS Number: 54827-17-7

Order Now

Specification Test Results

Abs.@450nm (50%, 1M H2SO4) ≤ 0.1
Abs.@650nm (undiluted) ≤ 0.1
Assay (P/F) PASS / FAIL
pH @25C (undiluted) 2.6 - 3.6

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR