Citric acid monohydrate ACS

Supplier: VWR
Warning

Synonyms: 2-Hydroxypropane-1,2,3-tricarboxylic acid, Hydroxytricarballylic acid, 2-Hydroxy-1,2,3-propanetricarboxilic acid

0908-2.5KG 0908-500G
97062-510CS 848.16 USD
97062-510 97062-512
Citric acid monohydrate ACS
Citric acid monohydrate
Formula: C₆H₈O₇.H₂O
Melting Pt: 135…152 °C
Density: 1.552 g/cm³ (20 °C)
Flash Pt: 173.9 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00149972
CAS Number: 5949-29-1
EINECS: 201-069-1
Merck Index: 13,02350
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3160681f96abd248f56672a96b72cc5f6e9d89a2&vid=c4db52c32c630d2c9876e60fc3e9e70a117ea0d8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Carbonizable Substances Pass
Chloride = 0.001%
Insolubles = 0.005%
Iron = 3 ppm
Lead = 2 ppm
Oxalate Test Pass
Phosphate = 0.001%
Purity 99% - 102%
Residue after Ignition = 0.02%
Sulfur Compounds (as SO4) = 0.002%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR