Riboflavine (vitamin B2), high_purity

Supplier: VWR

Synonyms: Vitamin B2, Lactoflavin, 7,8-Dimethyl-10-((2S,3S,4R)-2,3,4,5-tetrahydroxypentyl)benzo[g]pteridine-2,4(3H,10H)-dione, (-)-Riboflavin, Vitamine G

0744-250G
97062-302CS 579.14 USD
97062-302 97062-304
Riboflavine (vitamin B2), high_purity
Riboflavine (Vitamin B2)
Formula: C₁₇H₂₀N₄O₆
MW: 376.37 g/mol
Melting Pt: 280
Storage Temperature: Ambient
MDL Number: MFCD00005022
CAS Number: 83-88-5
EINECS: 201-507-1
Merck Index: 12,08367

Order Now

Specification Test Results

Ash = 0.3 %
Loss on Drying = 1.5 %
Lumiflavin (O.D.) = 0.025
Purity = 98 %
Specific Rotation (-135°) -( -115°)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR