Ethylene glycol, high purity

Supplier: VWR
Warning

Synonyms: Monoethylene glycol, Ethane-1,2-diol, 2-Hydroxyethanol, Ethylene alcohol, Ethanediol, Glycol alcohol, 1,2-Dihydroxy ethane, 1,2-Ethanediol, 1,2-Dihydroxyethane, Glycol, Dihydroxyethane

0582-1L 0582-4L
97061-964EA 60.43 USD
97061-964 97061-966
Ethylene glycol, high purity
Ethylene glycol

Colorless, slightly viscous liquid.Clear with no visible particulates.

Formula: HOCH₂CH₂OH
MW: 62.07 g/mol
Boiling Pt: 196…198 °C (1013 hPa)
Melting Pt: -13
Density: 1.115 g/cm³ (20 °C)
Flash Pt: 116
Storage Temperature: Ambient
MDL Number: MFCD00002885
CAS Number: 107-21-1
EINECS: 203-473-3
REACH: 01-2119456816-28
Merck Index: 13,03832
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4d1c97392851ba184f445f6ee912f84631f98d9&vid=2002300f174929a030d66d9f1394eb6bee89b7dc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Acidity = 0.002 %
Iron = 0.0005 %
Moisture (KF) = 0.5 %
Purity = 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR