AcroPrep™ Advance Filter Plates for Lysate Clearance, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
8275 8075 8175 8040
97052-132PK 233.37 USD
97052-132 97052-116 97052-130 97052-114
AcroPrep™ Advance Filter Plates for Lysate Clearance, Cytiva (Formerly Pall Lab)
Microplates Filter Plates
AcroPrep™ Advance Filter Plates for Lysate Clearance are manufactured with biologically inert materials that allow clarification of most types of lysates without loss of target molecules.
  • Biologically inert material
  • No loss of target molecules
  • Integrated prefilter
They have integrated prefilter that yields consistent filtration of highly particulated samples.
The AcroPrep Advance filter plates have a new well design that results in faster, more uniform filtration rates across the plate with reduced hold-up volume. The new outlet tip geometryMore Product Information