Portable Turbidity Meters, Turb Series, WTW
Supplier: WTW
600311 600312 600320 600326
97043-006EA 5535.9 USD
97043-006 97043-008 97043-010 97043-012
Portable Turbidity Meters, Turb Series, WTW
Turbidity Meters
The Turb® portable turbidity meters provide the user with the choice to perform nephelometric measurements at 90° scattered light with either a IR light source or tungsten white light source.
Small, lightweight, portable turbidity meters, typically used for spot analysis in the field - especially for all applications > 1NTU.
Battery power for more than 1500 measurements
Water proof housing (IP 67 compliance) with easy to use operation
Rugged carrying case for field use
The Turb 430 Series offers portable meters, with lab option via LabStation, for all applications includingMore Product Information