Benchtop pH/MV Meter, Sper Scientific

Supplier: Sper Scientific
860031
97039-648EA 1345.89 USD
97039-648
Benchtop pH/MV Meter, Sper Scientific
Electrochemistry Meters pH and Multi-Parameter Meters
Rugged housing with large LCD displaying the parameter being measured together with time, date and temperature (in °C or °F)

Operates on AC voltage (no batteries). Comes ready to use with ATC pH electrode, electrode holder arm, AC adaptor, computer cable and software. Also accepts any pH or ORP probe with a standard BNC connector.

Features automatic or manual temperature compensation, automatic or manual ranging, automatic buffer recognition, digital and analog outputs, min/max and 99 data points, data hold, 5 calibration points, and electrode status indication.

Ordering information: 5-year warranty.
Order Now

SPECIFICATIONS

Environmentally Preferable
Dimensions 217 × 168 × 58 mm (8.5 × 6.5 × 2.25")
Weight 533 g (18 oz.)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR