Advanced pH Meter Kit, Sper Scientific
Supplier: Sper Scientific
850056
97036-028EA 1087.57 USD
97036-028
Advanced pH Meter Kit, Sper Scientific
Electrochemistry Meters pH and Multi-Parameter Meters
Advanced pH Meter model 850055 reads either pH (in 0.01 resolution) or mV in large digits, with time, date, and temperature (in °C or °F) shown simultaneously on the lower display
Also indicates stable reading, low battery and displays calibration data. Scroll 99 data points with time stamps and min/max readings directly on the backlit LCD. Features hold, up to 5 point calibration, automatic buffer recognition, manual or automatic temperature compensation, and a USB port.
Ordering information: 5-year warranty. Kit includes Advanced pH Meter model 850055 in a hard carrying case completeMore Product Information