Purospher® STAR RP-18 Endcapped UHPLC Columns, MilliporeSigma
Supplier: MilliporeSigma
1.50645.0001 1.50646.0001 1.50650.0001 1.50648.0001 1.50651.0001 1.50653.0001 1.50654.0001 1.50649.0001
97021-978EA 622.54 USD
97021-978 97021-980 97021-982 97021-983 97021-984 97021-985 97021-986 EM1.50649.0001
Purospher® STAR RP-18 Endcapped UHPLC Columns, MilliporeSigma
Chromatography Columns
Available with optimised 2 μm particulate silica, these UHPLC columns based on Purospher® STAR RP-18 endcapped are ideal for ultra-fast applications, where resolution, sensitivity and sample throughput are crucial.
  • In UHPLC environment, speed is increased by a factor of 10
  • Solvent consumption is drastically cut
  • Excellent peak shape for all types of acidic, basic and metal chelating analytes
  • Extraordinary pH stability from pH 1.5 to 10.5 for an extremely wide application range
  • Enhanced sensitivity due to improved signal-to-noise ratio