Lumina™ Palladium (Pd) Hollow Cathode Lamp, PerkinElmer
Supplier: PerkinElmer
N3050158
97013-366EA 1942.85 USD
97013-366
Lumina™ Palladium (Pd) Hollow Cathode Lamp, PerkinElmer
Lights Hollow Cathode Lamps
Series N30501XX Lumina™ 50 mm (2") diameter single-element lamps are designed to be used with the PinAAcle™ and AAnalyst™ Atomic Absorption spectrometer series of instruments.
  • Single Element Lumina™ hollow cathode lamp (HCL) for the detection of elemental Palladium (Pd)
  • Lumina™ hollow cathode lamps may also be used with all earlier AA spectrometers that do not have automatic code reading capability by using an appropriate adapter cable
Every genuine PerkinElmer lamp is designed for use with and tested on spectrometers to assure compatibility and theMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable