Sapphire Injector (Int. Ø 2.0 mm) for Optima® 3x00 XL/3x00 DV/3000 SCX/4300V/5300V/7300V, PerkinElmer
Supplier: PerkinElmer
N0695494
97012-956EA 2437.59 USD
97012-956
Sapphire Injector (Int. Ø 2.0 mm) for Optima® 3x00 XL/3x00 DV/3000 SCX/4300V/5300V/7300V, PerkinElmer
Microinjectors Microinjectors, Programmable
This sapphire injector is for use with Optima® 3x00 XL/3x00 DV/3000 SCX/4300V/5300V/7300V ICP-OES series instruments. Sapphire injectors are ideal for silicon applications.
A Int. Ø 2.0 mm alumina injector (97012-836) can also be used for HF and aqua regia applications.