Pocket Digital Refractometer, Brix 0 to 88%, Sper Scientific
Supplier: Sper Scientific
300053
97011-842EA 1789.6 USD
97011-842
Pocket Digital Refractometer, Brix 0 to 88%, Sper Scientific
Refractometers Handheld Refractometers
Measure concentrations in brix, salinity, and a wide variety of other parameters in the field with laboratory accuracy
As small, light, and simple to use as optical refractometers, but with the added precision of a digital readout. Reads samples as small as 1ml. Sample temperature is initially displayed on the large LCD followed by the measurement results. The sophisticated microprocessor provides automatic calibration, automatic temperature compensation, and automatic shutoff (after 3 minutes). A sturdy water-resistant case protects the electronics from wet environments and allows forMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable