Swingsette™ M515 Tissue Processing/Embedding Cassettes, Simport
Supplier: Simport Scientific
M515-6 M515-12 M515-18 M515-3 M515-7 M515-11 M515-2 M515-4 M515-8 M515-17
97000-452CS 55.76 USD
97000-452 97000-454 97000-466 97000-472 97000-448 97000-456 97000-462 97000-464 97000-446 97000-450 97000-458 97000-470
Swingsette™ M515 Tissue Processing/Embedding Cassettes, Simport
Embedding Cassettes Tissue Processing Cassettes
Disposable plastic cassettes hold tissue specimens during processing, embedding, and storage
A special hinge holds the base and cover together and allows cassettes to be opened and closed as often as necessary. The cover can be removed and reinserted easily without the danger of specimen loss. Anterior surface is slanted at a 45° angle for use with leading labeling instruments. Three frosted writing surfaces. Cassette colors are non-cytotoxic and non-metallic.
They are molded from a special, high-density acetal polymer and are completely resistant to the chemical action of histologicalMore Product Information