Labtainer™ BioProcess Containers, 3 Ports, Thermo Scientific
Supplier: LIFE TECHNOLOGIES CORP
SH30713.02 SH30709.01 SH30709.03 SH30712.01 SH30712.02 SH30713.01 SH30713.05 SH30709.05 SH30714.02 SH30709.02 SH30709.04 SH30712.03 SH30712.04 SH30712.05 SH30713.03 SH30713.04 SH30714.01 SH30714.03 SH30714.04 SH30714.05
95059-576EA 102.02 USD
95059-576 95059-554 95059-558 95059-564 95059-566 95059-574 95059-582 95059-562 95059-586 95059-556 95059-560 95059-568 95059-570 95059-572 95059-578 95059-580 95059-584 95059-588 95059-590 95059-592
Labtainer™ BioProcess Containers, 3 Ports, Thermo Scientific
Bags Media Bags
Small volume liquid handling systems for cell culture and bioprocessing.
  • Space saving and ergonomic
  • Dispensing, packaging and storing cell culture media, buffers and process liquids
  • Delivery of cell culture media or process liquids to small scale bioproduction systems
  • Bioreactor and fermentation feed, sampling and harvest
  • Storage and transport of bulk intermediate products, process intermediates, vaccine conjugates and other biological products
They range in size from 50 ml to 50 L, with a variety of standard configurations to meet most applicationMore Product Information