HiTrap IMAC FF Affinity Chromatography Columns, Cytiva
Supplier: Cytiva
17-0921-04 17-0921-02
95055-958PK 805.69 USD
95055-958 95056-228
HiTrap IMAC FF Affinity Chromatography Columns, Cytiva
Chromatography Columns
HiTrap IMAC FF is prepacked with IMAC Sepharose 6 Fast Flow. The resin is charged with the metalion of your choice for Immobilized Metal ion Affinity Chromatography (IMAC) and subsequent purification of polyhistidine tagged proteins.
  • For optimizing purification of histidine-tagged proteins when Ni²⁺ is not the best choice of metal ion
  • Conveniently charge with your metal of choice
  • Optimal bead size for scaling up
  • High binding capacity
IMAC Sepharose FF is supplied free of metal ions. It is charged by the user with the transition metal ion of choice (e.More Product Information