HiTrap Q HP Anion Exchange Chromatography Columns
Supplier: Cytiva
17-1154-01 17-1153-01 29-0513-25
95056-078PK 561.93 USD
95056-078 95055-866 89501-392
HiTrap Q HP Anion Exchange Chromatography Columns
Chromatography Columns
HiTrap® Q HP are prepacked, ready-to-use High Performance strong anion exchange columns for high-resolution, small-scale protein purification.
  • Convenient and reproducible for fast, easy, high-performance separations either alone or connected in series
  • 34 µm bead size for high-performance, high-resolution purifications
  • Strong quaternary ammonium (Q) anion exchanger
  • Designed for use with a syringe, peristaltic pump, and chromatography systems such as ÄKTA design
The Q functional group is a quaternary amino group coupled to the matrix via chemicallyMore Product Information