Beef Extract, Certified, Dehydrated Culture Media, HiMedia
Supplier: HiMedia
CR002-500G
95041-190EA 260.53 USD
95041-190
Beef Extract, Certified, Dehydrated Culture Media, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
For the maximum recovery and growth of a wide variety of microorganisms
  • Microbiological culture media
  • Packaged in 500 g poly bottles
Usually incorporated in a concentration of 0.3% to 0.5% when combined with suitable peptones