Sodium sulfate, anhydrous ≥99% USP, Ph. Eur., BP suitable for use as excipient

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Disodium sulfate, Bisodium sulfate, Disodium sulphate, Sulfuric acid disodium salt, Bisodium sulphate, Sulphuric acid disodium salt

SO204-2.5KG SO204-12KG SO204-500GM
95035-970EA 582.05 USD
95035-970 700001-490 95035-972
Sodium sulfate, anhydrous ≥99% USP, Ph. Eur., BP suitable for use as excipient
Sodium sulphate

Sodium Sulfate, Anhydrous, USP, EP, BP is an electrolyte replenisher and is used in isosmotic solutions. All Spectrum Chemical USP, EP, BP productsare manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities

Formula: Na₂SO₄
Boiling Pt: 1429 °C (1013 hPa)
Melting Pt: 888 °C
Density: 2.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003504
CAS Number: 7757-82-6
EINECS: 231-820-9
Merck Index: 12,08829
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a4977cb8ca54638f22a60675bfbef71bbb77ab6&vid=942501aaa5afc8d85b82628854aac94d517322eb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR