Disodium hydrogen phosphate dihydrate ≥98% FCC suitable for use as excipient

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: sec-Sodium phosphate, Sodium monohydrogen phosphate dihydrate, Sodium hydrogenphosphate dihydrate, di-Sodium hydrogen orthophosphate dihydrate, Sodium phosphate dibasic dihydrate, Disodium phosphate, di-Sodium hydrogen phosphate dihydrate, Sorensen's salt

S1108-12KG S1108-45KGBL S1108-2.5KG S1108-500GM
700001-640EA 1063.9 USD
700001-640 700001-642 95035-894 95035-896
Disodium hydrogen phosphate dihydrate ≥98% FCC suitable for use as excipient
di-Sodium hydrogen phosphate dihydrate

Sodium Phosphate, Dibasic, Dihydrate, FCC is used in foods to adjust pH. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: Na₂HPO₄·2H₂O
Melting Pt: 92.5 °C
Density: 2.1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149182
CAS Number: 10028-24-7
EINECS: 231-448-7

Order Now

Specification Test Results

Assay 98% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR