Potassium benzoate ≥99% FCC suitable for use as excipient

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Benzoic acid potassium salt

PO156-2.5KG PO156-500GM
95035-084EA 299.45 USD
95035-084 95035-086
Potassium benzoate ≥99% FCC suitable for use as excipient
Potassium benzoate

Potassium Benzoate, FCC is used as a preservative that limits the growth of yeast, mold and some bacteria. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₇H₅KO₂
MW: 160.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00013061
CAS Number: 582-25-2
EINECS: 209-481-3

Order Now

Specification Test Results

Assay 99% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR