Methyl salicylate ≥98% FCC suitable for use as excipient

Supplier: SPECTRUM CHEMICAL MFG CORP
Warning

Synonyms: Wintergreen Oil, Methyl 2-hydroxybenzoate

M1363-4LT M1363-500ML
95034-444EA 699.5 USD
95034-444 95034-446
Methyl salicylate ≥98% FCC suitable for use as excipient
Methyl salicylate

Methyl Salicylate, FCC is used as a fragrant in food and beverages. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: 2-(HO)C₆H₄CO₂CH₃
Boiling Pt: 432 °C (1013 hPa)
Melting Pt: 100…101 °C
Density: 1.184
Flash Pt: 99 °C
Storage Temperature: Ambient
MDL Number: MFCD00002214
CAS Number: 119-36-8
EINECS: 204-317-7
Merck Index: 13,06143
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d636eb895c055579d494c4b8ed1636c32f052dd7&vid=7f7da4bc95980831012b381705f4b5a3f4ab7003&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 98% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR