Glycerine natural 99,0 Ph. Eur., USP, BP

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Glyceritol, Glysanin, 1,2,3-Propanetriol, Osmoglyn, Glycerin, Glycyl alcohol, Glyrol, Propane-1,2,3-triol, Propanetriol, Trihydroxypropane, Glycerol

G1016-200LT G1016-4LTPL G1016-200LTB G1016-20LTPL G1016-4GLCS4 G1016-4LTGL G1016-4PLCS4 G1016-500PC6 G1016-500GC6 G1016-500MLG G1016-500MLP G1016-500MLG G1016-500MLP G1016-1LT
89050-424EA 5030.25 USD
89050-424 95033-402 700000-216 700000-218 95033-398 95033-400 95033-404 95033-412 95033-406 700000-220 700000-222 95033-408 95033-410 75811-218
Glycerine natural 99,0 Ph. Eur., USP, BP
Glycerine

Glycerin, Natural, USP, EP, BP is one of two USP grade Glycerins available from Spectrum.


All Spectrum Chemical USP, EP, BP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 

Formula: HOCH₂CH(OH)CH₂OH
MW: 92.09 g/mol
Boiling Pt: 290
Melting Pt: 18.6
Density: 1.264
Flash Pt: 177
Storage Temperature: Ambient
MDL Number: MFCD00004722
CAS Number: 56-81-5
Merck Index: 13,04497
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3668cce85af1464db21fd098526252076b567956&vid=86a15453b54e7106c121d32e4e716385003c4507&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (Anhydrous Basis) 99.0 - 101.0%
Specific Gravity @ 25°C 1.249 min.
Appearance of Solution To Pass Test
Refractive Index 1.470 - 1.475
Identification To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR