Anti-CK7 Mouse Monoclonal Antibody [clone: OV-TL 12/30]

Supplier: Diagnostic Biosystems

MOB057-05 MOB057 PDM097
95024-456EA 360.28 USD
95024-456 99990-226 99990-974
Anti-CK7 Mouse Monoclonal Antibody [clone: OV-TL 12/30]
Antibodies
This antibody recognizes keratin polypeptide of 54 kDa. It stains a number of epithelial cells including ductal and glandular epithelia. Stratified squamous epithelia are positive but transitional epithelium of the urinary tract does not show any staining. This antibody labels adenocarcinomas and transitional carcinomas but does not react with squamous cell carcinomas.

Type:
Antigen: CK7
Clonality: Monoclonal
Clone: OV-TL 12/30
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR