Anti-AdV Mouse Monoclonal Antibody [clone: M58 + M73]
Supplier: Diagnostic Biosystems
MOB355 MOB355-05
99990-664EA 724.53 USD
99990-664 95024-222
Anti-AdV Mouse Monoclonal Antibody [clone: M58 + M73]
Antibodies
This antibody reacts with adenovirus. The adenovirus early gene product E1A is a potent stimulator of cellular proliferation, which when overexpressed can overcome the growth-inhibitory effects of TGF-β. The E1A region encodes a series of related proteins (35-46 kDa) with multifunctional capabilities and forms a specific complex with the retinoblastoma tumor suppressor gene product. The E1A and E1B regions together comprise the transforming region of adenovirus. While expression of E1A alone is sufficient to immortalize primary cells, complete transformation requires the additionalMore Product Information