Craig’s Medium Dehydrated Culture Media, HiMedia
Supplier: HiMedia
M974-500G
95022-572EA 323.7 USD
95022-572
Craig’s Medium Dehydrated Culture Media, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
For the cultivation of Vibrio cholerae to determine enterotoxigenicity.
  • Microbiological culture media
  • 500 g poly bottles