Milk Medium with Reducing Agent, Dehydrated, HiMedia Laboratories
Supplier: HiMedia
M816-500G
95022-298EA 222.61 USD
95022-298
Milk Medium with Reducing Agent, Dehydrated, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
For determination of the litmus reaction of Clostridium species.
  • Microbiological culture media
  • 500 g poly bottles