Enterococcus Confirmatory Agar Dehydrated Culture Media, HiMedia
Supplier: HiMedia
M392-500G
95021-766EA 238.57 USD
95021-766
Enterococcus Confirmatory Agar Dehydrated Culture Media, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
For confirming the presence of enterococci in water supplies and other sources.
  • Microbiological culture media
  • 500 g poly bottles