Ficoll-Paque® PLUS and Ficoll-Paque® PREMIUM Centrifugation Media, Cytiva
Supplier: Cytiva
17-5442-02 17544203 17-1440-02 17-5446-02 17-1440-03 17-5446-52
95021-207PK 287.36 USD
95021-207 95038-170 95021-205 95040-394 95038-168 95040-396
Ficoll-Paque® PLUS and Ficoll-Paque® PREMIUM Centrifugation Media, Cytiva
Cell Separation Media
Ficoll-Paque™ PREMIUM products are a range of sterile, ready to use density gradient media for the preparation of mononuclear cells.
  • Manufactured according to GMP and ISO 13485:2003 standards and the recommendations of the United States Pharmacopeia
  • Sterile and ready to use
  • Low levels of endotoxin (<0,12 EU/ml) secured and tested
  • Non cytotoxic
Sterile, ready to use reagents used to prepare mononuclear cell preparations from peripheral blood, bone marrow, and umbilical cord blood by density gradient centrifugation. Liquid physical state. ColorlessMore Product Information