MUG EC 0157 Agar Dehydrated Culture Media, HiMedia
Supplier: HiMedia
M1373-500G
95021-038EA 431.75 USD
95021-038
MUG EC 0157 Agar Dehydrated Culture Media, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
For the isolation and differentiation of enterohemorrhagic Escherichia coli O157:H7 from foodstuffs, water, and clinical samples by a fluorogenic method.
  • Microbiological culture media
  • 500 g poly bottles