T-Mop™ Cleanroom Mop Covers, Micronova
Supplier: Micronova
T6-14IR T6-18IR TAA-18IR T20-14IR T612-14IR T612-18IR QDST-14 QDST-18 T6-14 T6-18 QDT-14 T612-18 T612-14 T20-14 TAA12-14 TAA12-18 TAA-14 TAA-18 TMC6-14 JZTAA12-18 TAA-10 QPT-14 T11-14 QPT-18
43300-960CS 417.89 USD
43300-960 43300-962 43300-964 68400-596 89089-324 89089-340 89092-134 94000-438 94023-806 94023-808 68400-394 68400-396 68400-398 68400-404 68400-408 68400-854 89092-132 94023-866 94023-868 94023-874 94023-876 94023-878 94023-880 89401-702 10125-946 10146-120 10204-932 10142-777 76081-590 76247-298 76251-602
T-Mop™ Cleanroom Mop Covers, Micronova
Mops
A series of rounded T-mop covers with snaps that easily roll down walls, as well as clean awkward corners, tight spaces, and under equipment. Available in a range of fabrics for specialized cleaning.
  • Low profile, rounded covered sponges
  • Excellent on all room surfaces and equipment exteriors
  • Easily fits in awkward corners, tight spaces, and under equipment
  • Autoclavable
  • Holds up to strong disinfectants
Selection of fabrics include:
NovaPoly™ - 100% smooth knit polyester provides excellent disinfectant dispersal
PolySorb™ - 100% textured polyester forMore Product Information