Kapak 400 and 500 Series Tubular Rollstock, Ampac Flexibles
Supplier: Ampac Flexibles
TRS-16125-4 TRS-95125-3 TRS-95250-2 TRS-24250-6
11213-800CS 2302.71 USD
11213-800 11213-805 93001-502 10027-542
Kapak 400 and 500 Series Tubular Rollstock, Ampac Flexibles
Bags Roll Stock
Ampac’s polyester/polyethylene laminated tubular rollstock features the same properties as our laminated pouches. Since all tubular rollstock can be cut to any length,it allows for evidence of many different sizes to be collected and sealed for laboratory testing and long-term storage.
  • Excellent for containing accelerants
  • Low release of background volatiles
  • Hydrocarbon free
  • Prevents cross-contamination
Regular weight 2.5 mil material is best for moderate protection from handling and chemical or environmental contact. Tear notches included. Heavy-More Product Information