Dissolution Cannula Filters, Quality Lab Accessories
Supplier: QLA
FIL001-HR FIL004-HR FIL010-01-A FIL010-01 FIL010-DK FIL010-HR-A FIL010-HR FIL035-01-A FIL035-01 FIL035-HR FIL045-DK FIL070-01 FIL070-DK FIL070-HR FIL10S-DK FIL10S-HR FIL70S-HR FIL090-DK FILTIP-05 FILTIP-10 FILTIP-20 FILTIP-45 FIL045-01-A FIL070-EW-A FIL45S-DK FIL45S-DK-A FIL001-CA FIL001-CA-A FIL001-PT FIL005-PT FIL005-PT-A FIL010-CA FIL010-CA-A FIL010-PT FIL010-PT-A FIL010-VK FIL010-VK-A FIL020-01 FIL020-01-A FIL020-EW FIL020-EW-A FIL020-HR FIL020-HR-A FIL035-CA FIL035-CA-A FIL035-VK FIL035-VK-A FIL045-HR-A FIL070-VK FIL070-VK-A FIL20S-DK FIL20S-DK-A FIL45S-HR FIL45S-HR-A BSK020-DKC FIL001-PT-A FIL004-CA FIL004-PT FIL004-PT-A FIL035-PT-A FIL040-CA FIL040-CA-A FIL070-CA FIL070-CA-A FIL070-PT FIL070-PT-A FIL070-ST FIL070-ST-A FIL010-01K-A FIL010-ST-A FIL020-ST FIL035-ST-A FIL020-ST-A FIL20S-HR FIL001-EW-A FIL001-EW FIL004-EW-A FIL004-EW FIL010-EW-A FIL010-EW FIL035-EW FIL035-EW-A FIL040-EW-A FIL040-EW FIL045-01 FIL070-EW FIL10S-DK-A FIL35S-DK-A FIL35S-DK FIL35S-HR FIL35S-HR-A FIL70S-DK FIL70S-DK-A FILTIP-02 FIL10S-HR-A FIL001-HR-A FIL010-ST FIL035-ST FIL010-01K FIL004-HR-A FIL035-HR-A FIL070-01-A FIL070-HR-A FIL70S-HR-A FIL001-01 FIL001-01-A FIL002-01 FIL002-01-A FIL020-DK FIL020-DK-A FIL020-PT FIL020-PT-A FIL035-DK FIL035-DK-A FIL035-PT FIL045-DK-A FIL045-VK FIL045-VK-A FIL070-DK-A FIL090-DK-A FIL10S-ST FIL20S-HR-A
11010-252PK 1043 USD
11010-252 11010-256 11010-258 11010-260 11010-262 11010-264 11010-266 11010-268 11010-270 11010-274 11010-276 11010-280 11010-282 11010-286 11010-288 11010-292 11010-296 11010-298 11010-304 11010-306 11010-308 11010-310 89039-816 89039-818 89039-820 89039-822 89049-240 89049-242 89049-244 89049-246 89049-248 89049-250 89049-252 89049-254 89049-256 89049-258 89049-260 89049-262 89049-264 89049-266 89049-268 89049-270 89049-272 89049-274 89049-276 89049-278 89049-280 89049-284 89049-286 89049-288 89049-290 89049-292 89049-294 89049-296 89080-280 89080-294 89080-296 89080-300 89080-302 89080-306 89080-308 89080-310 89080-312 89080-314 89080-316 89080-318 89080-320 89080-322 89132-998 89133-002 89133-004 89133-012 89173-112 89173-114 93000-984 93000-986 93000-988 93000-990 93000-992 93000-994 93000-996 93000-998 93001-000 93001-002 93001-004 93001-006 93001-010 93001-012 93001-014 93001-016 93001-018 93001-020 93001-022 11010-302 11010-290 11010-250 89140-986 89140-992 89140-982 11010-254 11010-272 11010-278 11010-284 11010-294 75848-904 75848-906 75848-908 75848-910 75848-914 75848-916 75848-918 75848-920 75848-922 75848-924 75848-926 75848-928 75848-930 75848-932 75848-934 75848-936 75848-938 75848-940
Dissolution Cannula Filters, Quality Lab Accessories
Filters Filter Tubes
Only QLA manufactures pure quality filters which are guaranteed chromium and heavy metal free.
  • Made of the purest and highest quality UHMW Polyethylene (and PVDF where applicable)
  • All filters are made in the USA
  • Available in 100 filter packs or 1000 filter jars
Filters for most sampling methods are available in numerous micron ratings and three main types: cannula filters, filter discs, and filter tips. The PureQuality certification program guarantees filters not to have any chromium, heavy metals, material contaminants, or inorganic elementalMore Product Information