XPert™ Filtered Balance Stations, Labconco®
Supplier: Labconco
3940201 3940202 3940221 3940222 3940301 3940302 3940321 3940322 3940401 3940402 3940421 3940422 3940200 3940220 3940300 3940320 3940400 3940420 3946500 3946501 3946502 3946520 3946521 3946522 3946530 3946531 3946532 3946540 3946541 3946542 3946600 3946601 3946602 3946620 3946621 3946622 3946630 3946631 3946632 3946640 3946641 3946642
82018-132EA 8135 USD
82018-132 82018-134 82018-136 82018-138 82018-140 82018-142 82018-144 82018-146 82018-148 82018-150 82018-152 82018-154 89004-494 89004-496 89004-498 89004-500 89004-502 89004-504 93000-564 93000-566 93000-568 93000-570 93000-572 93000-574 93000-576 93000-578 93000-580 93000-582 93000-584 93000-586 93000-588 93000-590 93000-592 93000-594 93000-596 93000-598 93000-600 93000-602 93000-604 93000-606 93000-608 93000-610
XPert™ Filtered Balance Stations, Labconco®
Fume Hoods and Enclosures Balances Enclosures
These stations provide user protection by keeping powders and particulates contained during weighing procedures
The deep interiors easily accommodate large micro and analytical balances. The Clean-Sweep™ airfoil allows air to sweep the work surface for maximum containment. The perforated rear baffle promotes horizontal laminar airflow to maximize containment. The upper containment sash foil bleeds air into the enclosure to direct concentrations of contaminants away from the operator’s breathing zone. The upper dilution air supply introduces room air at the top of the sash to diluteMore Product Information