Antibiotics and Antimycotics, BD Diagnostics
Supplier: BD
230705 231213 231268 231290 231552 231591 231607 231628 231658 231664 231682 230733 230749 231263 231264 231274 231275 231278 231286 231590 231593 231606 231613 231629 231660 230725 230793 231040 231041 231042 231271 231562 231597 231633 231635 231657 231641 231651
90002-062EA 56.05 USD
90002-062 90002-070 90002-186 90002-214 90002-234 90002-250 90002-334 90002-364 90002-370 90002-398 90002-426 90002-476 90002-486 90002-504 90002-072 90002-074 90002-228 90002-230 90002-238 90002-240 90002-244 90002-246 90002-332 90002-362 90002-368 90002-374 90002-396 90002-408 90002-428 90002-480 90002-068 90002-078 90002-116 90002-118 90002-120 90002-236 90002-248 90002-340 90002-342 90002-382 90002-432 90002-436 90002-474 90002-482 90002-446 90002-464
Antibiotics and Antimycotics, BD Diagnostics
Microbiology Test Systems Antibiotic Tests
Ideal for the qualitative susceptibility testing of mycobacteria and related organisms.
  • Reliable
  • Convenient
  • Rapid method
This collection of media products is used for the evaluation of susceptibility of microorganisms to anti-microbial agents.
Packaging: Each test includes 50 susceptibility disks per cartridge.