Sodium dihydrogen phosphate monohydrate USP, Ph. Eur., FCC

Supplier: Macco Organiques S.R.O

Synonyms: Sodium phosphate monobasic monohydrate, Sodium dihydrogen orthophosphate monohydrate, Monosodium phosphate monohydrate, Sodium dihydrogenorthophosphate monohydrate, Sodium orthophosphate monobasic monohydrate

51115010120
89523-446EA 903 USD
89523-446
Sodium dihydrogen phosphate monohydrate USP, Ph. Eur., FCC
Sodium dihydrogen phosphate monohydrate

For excipient use only.


Certificates: HACCP, ISO 9001:2008, Kosher including for Passover days, HALAL.


Certifications: Certificates: HACCP, ISO 9001:2008, Kosher including for Passover days, HALAL.


Ordering information: Minimum order quantities may apply; contact your VWR representative for more information.

Formula: NaH₂PO₄·H₂O
MW: 137.99 g/mol
Melting Pt: 100
MDL Number: MFCD00149208
CAS Number: 10049-21-5
Merck Index: 13,08734
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4e3b73d9200cc18a9423c5b98ad4affbd4aca8a&vid=a46f80a2f4b075e731486341a7be9b01c444a160&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR