Trichloroacetic acid (TCA) ≥99.0% ACS

Supplier: VWR
Danger

Synonyms: TCA, Trichloroethanoic acid, Aceto-Caustin, TCA

0133-100G
89500-550CS 721.94 USD
89500-550 89500-552
Trichloroacetic acid (TCA) ≥99.0% ACS
Trichloroacetic acid (TCA)
Formula: Cl₃CCOOH
Boiling Pt: 196 °C (1013 hPa)
Melting Pt: 54…56 °C
Density: 1.63 g/cm³ (20 °C)
Flash Pt: 113 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00004177
CAS Number: 76-03-9
EINECS: 200-927-2
Merck Index: 13,09700
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae34130684af3f73eccad44eac686e325855232d&vid=b600bf2ba67ce275263e6172675e131d3f263837&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Chloride 0.002%
Clarity of Solution Pass
Heavy Metals = 0.002%
Insoluble Matter = 0.01%
Iron = 0.001%
Nitrate = 0.002%
Phosphate = 5 ppm
Purity = 99%
Residue after Ignition = 0.03%
Substances Darkened by H2SO4 Pass
Sulfate = 0.02%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR