Stainless Steel Base Molds, Simport Scientific
Supplier: Simport Scientific
M474-1 M474-2 M474-3 M474-4 M474-5
89498-708PK 192.72 USD
89498-708 89498-710 89498-712 89498-714 89498-716
Stainless Steel Base Molds, Simport Scientific
Moulds Moulds for Histology
The M474 Stainless Steel Base Molds fit most cassettes and offer superior thermal exchange.
  • Manufactured from high-quality stainless steel
  • Optimal thermal conductivity
  • Highly polished surface for easy paraffin block removal
  • Well corners are rounded for optimal paraffin ribboning and a reusable design
Available in five sizes.
Packaging: Molds are supplied in a pack of 12.