Lab-Tip® 125S Cleanroom Swab, Berkshire
Supplier: Berkshire
LTP125S.25
89496-568CS 237.89 USD
89496-568
Lab-Tip® 125S Cleanroom Swab, Berkshire
Swabs Swabs for Cleaning
The Lab-Tip® 125S Large Sampling Swab is a cleanroom-laundered, double-layered polyester knit swab.
  • Autoclavable
  • Low particles, fibers, ions, and extractables
  • Excellent chemical resistance
  • Soft and non-abrasive with no contaminating adhesives
  • Good sorbency
  • Free from silicone, amides, and DOP
  • Cleanroom Class 100 to 100000
Grooves on the handle make for easy snapping. The cleanroom-laundered knitted head provides ultra-low NVR and ion levels. Contamination control, with critically low particles, fibers, ions, and extractables. Excellent chemicalMore Product Information