VWR® Cleanroom Sponges
Supplier: VWR International
VWSPAW-73 VWSPAW-73IR VWSP7-73 VWSP7-73IR VWSP68-73
89495-454CS 96.86 USD
89495-454 89495-456 89495-458 89495-460 89495-462 89495-464
VWR® Cleanroom Sponges
Wipes Sponges
A range of sponges, made with cleanroom materials, are designed for cleaning small parts, equipment, and other items in production or the lab.
  • Durable, versatile, and absorbent range of sponges
  • Hold up to alcohols, common cleaning solutions, and strong disinfectants
  • Reusable and autoclavable
Fabric cover options include a 100% polyester braided mesh, a cleanroom alternative to abrasive pads to remove residues, a 100% polyester knit, FDA approved for indirect food contact, and a white microfiber 80% polyester/20% polyamide split fiber fabric with highMore Product Information