Eurostar 200 Control Electronic Overhead Stirrer
Supplier: IKA Works
0003992001
89412-510EA 7283.8 USD
89412-510
Eurostar 200 Control Electronic Overhead Stirrer
Stirrers Overhead Stirrers
Extremely powerful laboratory stirrer for highly viscous applications and intensive mixing for quantities up to 100 L (H₂O).
  • Infinitely adjustable speed
  • Push-through agitator shafts
  • Overload protection, short-term overload operation
  • Slim casing, quiet operation
  • Rrror code display
The Eurostar 200 Control Electronic Overhead Stirrer features multilingual TFT display, programmable functions, integrated temperature measurement, interval operation, timer function, adjustabel safety circuit, locked function.
The unit is designed with a removableMore Product Information