Grant bio FTA-1 Portable Aspirator, Grant Instruments
Supplier: GRANT USA INC.
FTA-1
89402-922EA 2325.44 USD
89402-922
Grant bio FTA-1 Portable Aspirator, Grant Instruments
Pumps Vacuum Aspiration Systems
The aspirator with trap flask FTA-1 with a 1L trap flask is designed for the aspiration or removal of any remaining alcohol or buffer from microtest tube walls during DNA/RNA purification and other macromolecule re-precipitation techniques
  • All in one system with integrated pump
  • Trap flask volume 1L
  • Fitted with hydrophobic microbiological filter
  • Vacuum pressure of -500mbar
  • Small, compact and easy to use
  • Perfect for small volume aspiration
Compact, efficient, easy to use. Flexible for the user by offering a choice of either a single tip operation or aMore Product Information