TouchNTuff® DermaShield® 73-701 Cleanroom gloves, polychloroprene, sterile, Ansell
Supplier: Ansell Healthcare
387068 387069 387070 387071 387072 387073 387074
89402-798CS 965.67 USD
89402-798 89402-800 89402-802 89402-804 89402-806 89402-808 89402-810
TouchNTuff® DermaShield® 73-701 Cleanroom gloves, polychloroprene, sterile, Ansell
Gloves
Dermashield is made from a proprietary poly-chloroprene blend that is free of latex proteins and accelerators.
  • Cleanroom compatible
  • Straight cuffs
  • Excellent resistance to many chemicals
These sterile gloves are the ideal choice in the prevention of both Type 1 and Type IV allergies associated with natural rubber.
Dermashield 73-701 is clean and sterile and ideally suited for ISO class 5 (Class 100) enviroments. Neoprene composition offers chemical resistance to a broad spectrum of chemicals without compromising comfort and flexibility.