Calcium hydroxide, Technical Grade

Supplier: VWR
Danger

Synonyms: Slaked lime

N883-500G
89399-872EA 90.5 USD
89399-872 89399-874
Calcium hydroxide, Technical Grade
Calcium hydroxide
Formula: Ca(OH)₂
MW: 74.09 g/mol
Melting Pt: 550
Density: 2.24 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010901
CAS Number: 1305-62-0
EINECS: 215-137-3
UN: 3262
Merck Index: 13,01675

Order Now

Specification Test Results

Calcium Carbonate {CaCO3} 15%
Calcium Hydroxide {Ca(OH)2} 83 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR