VWR® Blood Bank Saline Solutions
Supplier: VWR International
89370-096 89370-098 89370-100 89370-102
89370-096EA 44.89 USD
89370-096 89370-098 89370-100 89370-102
VWR® Blood Bank Saline Solutions
Buffers Cell Culture Buffers
Blood bank saline is pH-controlled for a pH between 6.5 and 7.0.
  • pH-controlled
  • Enhances weak agglutination reactions
  • Stable without preservatives
Buffered saline is tightly controlled to a pH range of 7.0 to 7.2 and is specially designed to enhance weak agglutination reactions in blood bank procedures. Both saline and buffered saline remain stable without preservatives.
Order Now
SPECIFICATIONS
Environmentally Preferable