HiTrap Columns, Protein L Columns Prepacked with Capto L, Cytiva
Supplier: Cytiva
17547815 17547851 17-5478-55 29-0486-65
89234-992EA 1146.52 USD
89234-992 89234-994 89234-996 89501-382
HiTrap Columns, Protein L Columns Prepacked with Capto L, Cytiva
Chromatography Columns
Columns prepacked with Capto™ L, an affinity chromatography medium (resin) for capture of antibodies and antibody fragments such as Fab fragments, scFv, and Dabs.
  • Prepacked for easy screening and process development, as well as small scale purification
  • Columns easily connected in series for increased capacity
  • Designed for use with syringe, peristalic pump, or chromatography system