Protexis™ Latex Micro Sterile Latex Powder-Free Surgical Gloves with Nitrile Coating, Cardinal Health
Supplier: Cardinal Health
2D72NT55X 2D72NT60X 2D72NT65X 2D72NT70X 2D72NT75X 2D72NT80X 2D72NT85X 2D72NT90X
89233-814CS 1876.01 USD
89233-814 89233-816 89233-818 89233-820 89233-822 89233-824 89233-826 89233-828
Protexis™ Latex Micro Sterile Latex Powder-Free Surgical Gloves with Nitrile Coating, Cardinal Health
Gloves
Surgical, examination gloves provide additional protection.
  • Natural rubber latex formulation
  • Powder-free
  • 20% thicker than standard gloves
  • Triple-dip coated
  • Proprietary interlocking cuff style
These powder-free gloves help to reduce powder-related skin irritation and post-operative complications associate with granulomas. 20% thinner than Protexis™ Latex Surgical Gloves for enhanced tactile sensitivity. Triple-dip coating technology integrates nitrile and latex for effortless donning and glove strength. Excellent choice as an outer glove when double-More Product Information