Combitips® advanced Positive Displacement Pipette Tips, Sterile - Individually Wrapped
Supplier: Eppendorf
0030089510 0030089529 0030089537 0030089545 0030089553 0030089561 0030089570 0030089588 0030089596
89232-948CS 220.18 USD
89232-948 89232-950 89232-952 89232-954 89232-956 89232-958 89232-960 89232-962 89232-964
Combitips® advanced Positive Displacement Pipette Tips, Sterile - Individually Wrapped
Pipette Tips
Eppendorf Combitips® advanced, individually wrapped in 'Sterile' grade, offer exceptional purity and precision for sensitive lab procedures. Ideal for contamination-sensitive environments, these tips ensure accurate, sterile pipetting with ergonomic ease.
  • Combitips® advanced tips are the dedicated consumable for all Repeater®/Multipette® multi-dispenser pipettes from Eppendorf
  • 9 volume sizes (0.1 to 50 ml) offer a maximum range of dispensing volumes with increments as low as 100 nl
  • Combitips® advanced with the purity grade 'sterile' conform to a Sterility
More Product Information