ESP™ Pipette Tip Reloading Systems, Neptune™
Supplier: Biotix
63302159 63302166 63300538 63302162 63300523 63300560 63302179 63300570 63302176 63302170 63300511 63300801 63300802 63300803 63300804 63300800
89140-104CS 315.73 USD
89140-104 89140-900 89140-902 89168-738 89168-742 89168-748 89168-768 89168-772 89217-464 89425-640 89425-657 76311-242 76311-244 76311-246 76311-248 76311-290
ESP™ Pipette Tip Reloading Systems, Neptune™
Pipette Tips
Revolutionary ESP reload system offers the environmentally friendly reuse of existing trays.
  • Autoclavable
  • Good sustainability
  • Superior stability, firm placement
  • Maximum space saving design
Eco-friendly empty pipette tips trays are manufactured from recycled polypropylene. When combined with ESP™ reload systems, these tip trays reduce plastic waste by up to 90%.
Unique transfer system minimizes plastic waste by as much as 90%. Reload 10 trays in 90 seconds. Integrated transfer card prevents contamination by minimizing handling. Universal pipetteMore Product Information